siRNA Details
| Gene Symbol: Olfr159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019476.1 | NP_062349.1 | 1 | 488 | ||
| GenBank mRNA | AY043305.1 | - | 1 | 105 | ||
| BC113135.1 | - | 1 | 487 | |||
| BC134348.1 | - | 1 | 487 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr159| Gene Symbol: Olfr159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019476.1 | NP_062349.1 | 1 | 488 | ||
| GenBank mRNA | AY043305.1 | - | 1 | 105 | ||
| BC113135.1 | - | 1 | 487 | |||
| BC134348.1 | - | 1 | 487 | |||