siRNA Details
| Gene Symbol: Cxcl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019494.1 | NP_062367.1 | 4 | 1340 | ||
| GenBank mRNA | AF179872.1 | - | 4 | 1340 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcl11| Gene Symbol: Cxcl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019494.1 | NP_062367.1 | 4 | 1340 | ||
| GenBank mRNA | AF179872.1 | - | 4 | 1340 | ||