siRNA Details
| Gene Symbol: Dgke | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019505.3 | NP_062378.1 | 3, 4 | 1106 | ||
| GenBank mRNA | AF136744.1 | - | 3, 4 | 743 | ||
| AK044596.1 | - | 1842 | ||||
| AK049335.1 | - | 3, 4 | 906 | |||
| AK147841.1 | - | 3, 4 | 1107 | |||
| BB635763.1 | - | 3, 4 | 510 | |||

Dgke
Genomic Map