siRNA Details
| Gene Symbol: Fscn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019569.2 | NP_062515.2 | 2 | 651 | ||
| GenBank mRNA | AF176024.2 | - | 2 | 763 | ||
| BC050759.1 | - | 2 | 651 | 6770861 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fscn3| Gene Symbol: Fscn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019569.2 | NP_062515.2 | 2 | 651 | ||
| GenBank mRNA | AF176024.2 | - | 2 | 763 | ||
| BC050759.1 | - | 2 | 651 | 6770861 | ||