siRNA Details
| Gene Symbol: Tspan7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019634.2 | NP_062608.2 | 8 | 832 | ||
| GenBank mRNA | AF052492.1 | - | 8 | 780 | ||
| AK004776.1 | - | 8 | 792 | |||
| AK147306.1 | - | 7 | 5853 | |||
| AK147404.1 | - | 6 | 6124 | |||
| BC026516.1 | - | 7 | 631 | 4911317 | ||
| BC050153.1 | - | 8 | 780 | |||
| BC058603.1 | - | 8 | 788 | 6529011 | ||
| D26483.1 | - | 8 | 780 | |||

Tspan7
Genomic Map