siRNA Details
| Gene Symbol: Fam60a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019643.3 | NP_062617.2 | 2 | 210 | ||
| GenBank mRNA | AK046519.1 | - | 2 | 117 | ||
| AK145384.1 | - | 2 | 221 | |||
| AK166569.1 | - | 2 | 216 | |||
| BC076599.1 | - | 2 | 210 | 30614283 | ||
| U64033.1 | - | 2 | 299 | |||

Fam60a
Genomic Map