siRNA Details
| Gene Symbol: Wsb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019653.3 | NP_062627.3 | 2 | 471 | ||
| GenBank mRNA | AF033186.1 | - | 2 | 168 | ||
| AK019206.2 | - | 2 | 448 | |||
| AK145974.1 | - | 2 | 450 | |||
| AK146752.1 | - | 2 | 446 | |||
| AK150394.1 | - | 2 | 446 | |||
| AK150600.1 | - | 2 | 446 | |||
| AK152545.1 | - | 2 | 449 | |||
| AK159905.1 | - | 2 | 446 | |||
| AK167174.1 | - | 2 | 448 | |||
| AK167394.1 | - | 2 | 449 | |||
| AK167942.1 | - | 2 | 446 | |||
| AK168468.1 | - | 2 | 449 | |||
| AK168565.1 | - | 2 | 449 | |||
| AK169058.1 | - | 2 | 446 | |||
| BC053391.1 | - | 2 | 222 | 30073116 | ||

Wsb1
Genomic Map