siRNA Details
| Gene Symbol: Ppp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019674.3 | NP_062648.1 | 2, 3 | 215 | ||
| GenBank mRNA | AF088911.1 | - | 2, 3 | 240 | ||
| AK144244.1 | - | 1, 2 | 142 | |||
| BC001993.1 | - | 2, 3 | 215 | 3491568 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp4c| Gene Symbol: Ppp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019674.3 | NP_062648.1 | 2, 3 | 215 | ||
| GenBank mRNA | AF088911.1 | - | 2, 3 | 240 | ||
| AK144244.1 | - | 1, 2 | 142 | |||
| BC001993.1 | - | 2, 3 | 215 | 3491568 | ||