siRNA Details
| Gene Symbol: Cyb561d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019720.4 | NP_062694.1 | 3 | 436 | ||
| GenBank mRNA | AF131206.1 | - | 4 | 308 | ||
| AK030225.1 | - | 3 | 437 | |||
| AK162586.1 | - | 3 | 423 | |||
| BC027212.1 | - | 4 | 305 | 3993231 | ||

Cyb561d2
Genomic Map