siRNA Details
| Gene Symbol: Slc22a21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019723.2 | NP_062697.1 | 6 | 1209 | ||
| GenBank mRNA | AB018436.1 | - | 6 | 1209 | ||
| AK080177.1 | - | 5 | 1412 | |||
| AK133431.1 | - | 6 | 1138 | |||
| AK161508.1 | - | 6 | 1105 | |||
| BC132147.1 | - | 6 | 1172 | 40130424 | ||
| BC137908.1 | - | 6 | 1172 | 8860927 | ||

Slc22a21
Genomic Map