siRNA Details
| Gene Symbol: Stam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019667.2 | NP_062641.1 | 3, 4 | 249 | ||
| GenBank mRNA | AB012611.1 | - | 3, 4 | 264 | ||
| AK004604.2 | - | 3, 4 | 249 | |||
| AK035789.1 | - | 302 | ||||
| AK078244.1 | - | 3, 4 | 281 | |||
| AK159699.1 | - | 3, 4 | 278 | |||
| AK168729.1 | - | 3, 4 | 281 | |||
| BC013818.1 | - | 3, 4 | 285 | 3671426 | ||
| DQ141211.1 | - | 4, 5 | 479 | |||

Stam2
Genomic Map