siRNA Details
| Gene Symbol: Arid3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019689.2 | NP_062663.1 | 1 | 371 | ||
| GenBank mRNA | AF116847.1 | - | 1 | 371 | ||
| AK038068.1 | - | 2 | 452 | |||
| AK148073.1 | - | 2 | 468 | |||
| BC049776.1 | - | 2 | 593 | |||
| BC145330.1 | - | 1 | 365 | |||
| BC150755.1 | - | 1 | 365 | 9087666 | ||

Arid3b
Genomic Map