siRNA Details
| Gene Symbol: Clcnkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019701.2 | NP_062675.2 | 14 | 1382 | ||
| GenBank mRNA | AF124847.1 | - | 14 | 1375 | ||
| AK146876.1 | - | 13 | 1445 | |||
| BC019983.1 | - | 14 | 1408 | 4188062 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clcnkb| Gene Symbol: Clcnkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019701.2 | NP_062675.2 | 14 | 1382 | ||
| GenBank mRNA | AF124847.1 | - | 14 | 1375 | ||
| AK146876.1 | - | 13 | 1445 | |||
| BC019983.1 | - | 14 | 1408 | 4188062 | ||