siRNA Details
| Gene Symbol: Hbs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042593.1 | NP_001036058.1 | 4 | 538 | ||
| NM_001145209.1 | NP_001138681.1 | 4 | 538 | |||
| NM_019702.2 | NP_062676.2 | 4 | 538 | |||
| GenBank mRNA | AF087672.1 | - | 1 | 396 | ||
| AK006626.1 | - | 4 | 511 | |||
| AK012856.1 | - | 4 | 488 | |||
| AK146678.1 | - | 4 | 504 | |||
| AK166140.1 | - | 4 | 501 | |||
| AK168671.1 | - | 4 | 501 | |||
| AK169281.1 | - | 4 | 501 | |||
| AK173091.1 | - | 4 | 484 | |||
| BC010251.1 | - | 4 | 463 | 3964560 | ||
| BC085235.1 | - | 4 | 480 | 5328213 | ||
| BU510509.1 | - | 4 | 490 | |||

Hbs1l
Genomic Map