siRNA Details
| Gene Symbol: Nupr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019738.1 | NP_062712.1 | 2 | 235 | ||
| GenBank mRNA | AF131196.1 | - | 2 | 235 | ||
| AK009572.1 | - | 2 | 255 | |||
| BC002109.1 | - | 2 | 241 | 3487434 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nupr1| Gene Symbol: Nupr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019738.1 | NP_062712.1 | 2 | 235 | ||
| GenBank mRNA | AF131196.1 | - | 2 | 235 | ||
| AK009572.1 | - | 2 | 255 | |||
| BC002109.1 | - | 2 | 241 | 3487434 | ||