siRNA Details
| Gene Symbol: Stmn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019641.4 | NP_062615.1 | 5 | 574 | ||
| GenBank mRNA | AB064953.1 | - | 5 | 527 | ||
| AK002606.1 | - | 5 | 496 | |||
| AK012231.1 | - | 5 | 496 | |||
| AK012347.1 | - | 5 | 496 | |||
| AK013302.1 | - | 5 | 496 | |||
| AK014108.1 | - | 5 | 497 | |||
| AK088251.1 | - | 5 | 497 | |||
| AK088628.1 | - | 5 | 496 | |||
| BC010581.1 | - | 5 | 483 | 3154091 | ||
| BC031831.1 | - | 5 | 482 | 4500926 | ||
| BC054396.1 | - | 4 | 475 | 6492045 | ||
| X94915.1 | - | 3 | 404 | |||
| Gene Symbol: Gm11223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030082.1 | - | 571 | |||
| Gene Symbol: Stmn1-rs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CF896624.1 | - | 1 | 254 | ||
| CF897466.1 | - | 1 | 224 | |||

Stmn1
Genomic Map