siRNA Details
| Gene Symbol: Pias1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019663.3 | NP_062637.2 | 14 | 3009 | ||
| GenBank mRNA | AK028413.1 | - | 1 | 591 | ||
| AK034970.1 | - | 1 | 940 | |||
| AK075708.1 | - | 14 | 3010 | |||
| AK136188.1 | - | 3 | 459 | |||
| AK142088.1 | - | 1 | 918 | |||
| AK162584.1 | - | 1 | 940 | |||

Pias1
Genomic Map