siRNA Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019670.1 | NP_062644.1 | 12 | 1247 | ||
| GenBank mRNA | AF094519.1 | - | 12 | 1247 | ||
| AK137073.1 | - | 11 | 1336 | |||
| AK138694.1 | - | 12 | 1277 | |||
| AK161737.1 | - | 11 | 1265 | |||
| AK166882.1 | - | 12 | 1303 | |||
| AK220493.1 | - | 12 | 1311 | |||
| BC003250.1 | - | 11 | 1281 | |||
| BC028920.1 | - | 5 | 548 | 4008236 | ||
| BC085191.1 | - | 12 | 1357 | 30693873 | ||
| BC086779.1 | - | 12 | 1322 | |||

Diap3
Genomic Map