siRNA Details
| Gene Symbol: Fads2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019699.1 | NP_062673.1 | 12 | 1498 | ||
| GenBank mRNA | AF126798.1 | - | 12 | 1498 | ||
| AK083282.1 | - | 1465 | ||||
| AK143524.1 | - | 1516 | ||||
| AK185613.1 | - | 2 | 248 | |||
| BC057189.1 | - | 12 | 1486 | 5137343 | ||

Fads2
Genomic Map