siRNA Details
| Gene Symbol: Nme4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019731.1 | NP_062705.1 | 5 | 614 | ||
| GenBank mRNA | AF153451.1 | - | 5 | 614 | ||
| AF288690.1 | - | 5 | 627 | |||
| AK012805.1 | - | 5 | 650 | |||
| AK017724.1 | - | 5 | 630 | |||
| BC027277.1 | - | 5 | 632 | 3591383 | ||

Nme4
Genomic Map