siRNA Details
Gene Symbol: Tusc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019742.4 | NP_062716.1 | 3 | 1622 | ||
GenBank mRNA | AF123387.1 | - | 3 | 1509 | ||
AK082684.1 | - | 3 | 1623 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tusc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019742.4 | NP_062716.1 | 3 | 1622 | ||
GenBank mRNA | AF123387.1 | - | 3 | 1509 | ||
AK082684.1 | - | 3 | 1623 |