siRNA Details
| Gene Symbol: Dpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019759.2 | NP_062733.1 | 1 | 101 | ||
| GenBank mRNA | AF143374.1 | - | 1 | 101 | ||
| AK019890.1 | - | 1 | 190 | |||
| BC046420.1 | - | 1 | 127 | 4165966 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpt| Gene Symbol: Dpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019759.2 | NP_062733.1 | 1 | 101 | ||
| GenBank mRNA | AF143374.1 | - | 1 | 101 | ||
| AK019890.1 | - | 1 | 190 | |||
| BC046420.1 | - | 1 | 127 | 4165966 | ||