siRNA Details
| Gene Symbol: KCNC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004976.4 | NP_004967.1 | 2 | 1567 | ||
| GenBank mRNA | BC107129.2 | - | 2 | 1584 | 40024733 | |
| DA495469.1 | - | 1 | 92 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNC1| Gene Symbol: KCNC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004976.4 | NP_004967.1 | 2 | 1567 | ||
| GenBank mRNA | BC107129.2 | - | 2 | 1584 | 40024733 | |
| DA495469.1 | - | 1 | 92 | |||