siRNA Details
Gene Symbol: Akap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB028920.1 | - | 4 | 188 | ||
AK004801.1 | - | 4 | 194 | |||
AK030964.1 | - | 1 | 828 | |||
AK089092.1 | - | 4 | 201 | |||
AK143300.1 | - | 2 | 1576 | |||
BC042602.1 | - | 4 | 184 | |||
BC060170.1 | - | 4 | 179 | |||
BC060209.1 | - | 4 | 285 | |||
BC078448.1 | - | 4 | 172 | |||
BC125521.1 | - | 4 | 184 | 40130036 | ||
BC137746.1 | - | 4 | 184 | 8860765 |
