siRNA Details
| Gene Symbol: Snd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019776.2 | NP_062750.2 | 2 | 365 | ||
| GenBank mRNA | AB021491.1 | - | 2 | 322 | ||
| AK034914.1 | - | 2 | 313 | |||
| AK088028.1 | - | 2 | 324 | |||
| AK133837.1 | - | 2 | 371 | |||
| AK159837.1 | - | 2 | 330 | |||
| AK161591.1 | - | 2 | 319 | |||
| AK162432.1 | - | 2 | 306 | |||
| AK167576.1 | - | 2 | 355 | |||
| AK204079.1 | - | 1 | 31 | |||
| BB613775.1 | - | 2 | 365 | |||
| BC007126.1 | - | 2 | 312 | 3592548 | ||
| BM944959.1 | - | 1 | 43 | |||

Snd1
Genomic Map