siRNA Details
| Gene Symbol: Bloc1s6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019788.3 | NP_062762.1 | 2 | 235 | ||
| GenBank mRNA | AK038381.1 | - | 2 | 213 | ||
| AK077720.1 | - | 2 | 217 | |||
| AK147116.1 | - | 2 | 238 | |||
| AK160580.1 | - | 2 | 205 | |||
| AK170964.1 | - | 2 | 166 | |||
| AK172571.1 | - | 2 | 216 | |||
| BC016554.1 | - | 2 | 218 | |||
| BC085260.1 | - | 2 | 204 | 6437935 | ||
| BY015259.1 | - | 2 | 236 | |||

Bloc1s6
Genomic Map