siRNA Details
| Gene Symbol: Ube2g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019803.3 | NP_062777.2 | 5 | 350 | ||
| GenBank mRNA | AF296657.1 | - | 5 | 331 | ||
| AK136365.1 | - | 5 | 337 | |||
| AK146554.1 | - | 5 | 355 | |||
| AK154464.1 | - | 5 | 371 | |||
| AK178687.1 | - | 2 | 133 | |||
| BC010321.1 | - | 5 | 350 | 2651046 | ||
| U93241.1 | - | 4 | 250 | |||

Ube2g2
Genomic Map