siRNA Details
| Gene Symbol: Naa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177965.1 | NP_001171436.1 | 3 | 314 | ||
| NM_019870.3 | NP_063923.1 | 3 | 314 | |||
| GenBank mRNA | AJ627039.1 | - | 2 | 133 | ||
| AK003806.1 | - | 3 | 251 | |||
| AK009697.1 | - | 3 | 225 | |||
| AK014469.1 | - | 3 | 243 | |||
| AK078700.1 | - | 3 | 277 | |||
| AY183136.1 | - | 3 | 283 | |||
| AY183137.1 | - | 3 | 283 | |||
| BC027219.1 | - | 3 | 215 | 3673739 | ||
| BY101790.1 | - | 3 | 315 | |||
| Gene Symbol: Arhgap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK129060.1 | - | 201 | |||

Naa10
Genomic Map