siRNA Details
| Gene Symbol: Htra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019752.3 | NP_062726.3 | 2 | 703 | ||
| GenBank mRNA | AF164513.1 | - | 2 | 582 | ||
| AF175324.1 | - | 2 | 599 | |||
| AK011536.1 | - | 1509 | ||||
| AK146338.1 | - | 2 | 594 | |||
| AK159188.1 | - | 2 | 523 | |||
| AK184043.1 | - | 1 | 40 | |||
| BC033276.1 | - | 2 | 563 | |||
| BC049880.1 | - | 2 | 703 | 5321861 | ||

Htra2
Genomic Map