siRNA Details
| Gene Symbol: 1110004F10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019772.2 | NP_062746.1 | 2, 3 | 233 | ||
| GenBank mRNA | AB025407.1 | - | 2, 3 | 233 | ||
| AK003415.1 | - | 2, 3 | 274 | |||
| AK144059.1 | - | 2, 3 | 260 | |||
| AK167571.1 | - | 2, 3 | 262 | |||
| AK168206.1 | - | 2, 3 | 267 | |||
| AK169309.1 | - | 2, 3 | 260 | |||
| AK180958.1 | - | 2, 3 | 111 | |||
| BC072575.1 | - | 2, 3 | 269 | 5715413 | ||

1110004F10Rik
Genomic Map