siRNA Details
| Gene Symbol: Zbtb20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285805.1 | NP_001272734.1 | 10 | 1627 | ||
| NM_019778.2 | NP_062752.2 | 8 | 1574 | |||
| NM_181058.1 | NP_851401.1 | 6 | 1352 | |||
| GenBank mRNA | AB221632.1 | - | 2 | 1039 | ||
| AF185576.1 | - | 6 | 1397 | |||
| AF194030.1 | - | 4 | 1189 | |||
| AK005028.2 | - | 6 | 1353 | |||
| AK034574.1 | - | 1 | 226 | |||
| AY028963.1 | - | 3 | 1156 | |||
| BC031114.1 | - | 7 | 1460 | 4971586 | ||
| BC056446.1 | - | 10 | 1599 | 5703319 | ||

Zbtb20
Genomic Map