siRNA Details
| Gene Symbol: Ggcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019802.5 | NP_062776.1 | 10 | 1421 | ||
| GenBank mRNA | AF087938.1 | - | 10 | 1396 | ||
| AK033496.1 | - | 10 | 1348 | |||
| AK036577.1 | - | 10 | 1395 | |||
| AK135425.1 | - | 10 | 1408 | |||
| AK165982.1 | - | 10 | 1396 | |||
| BC138763.1 | - | 10 | 1346 | 8861785 | ||
| BC138764.1 | - | 10 | 1346 | 8861786 | ||

Ggcx
Genomic Map