siRNA Details
| Gene Symbol: Gkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019832.3 | NP_062806.1 | 2 | 354 | ||
| GenBank mRNA | AB033130.1 | - | 1 | 213 | ||
| AK016588.1 | - | 1 | 126 | |||
| AK140052.1 | - | 1 | 774 | |||
| BC052347.1 | - | 2 | 354 | 6774715 | ||

Gkap1
Genomic Map