siRNA Details
| Gene Symbol: Fam69b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019833.3 | NP_062807.2 | 4, 5 | 671 | ||
| GenBank mRNA | AB030186.1 | - | 4, 5 | 666 | ||
| AF332189.1 | - | 4, 5 | 665 | |||
| AK045880.1 | - | 4, 5 | 672 | |||
| AK050584.1 | - | 4, 5 | 585 | |||
| AK171112.1 | - | 3, 4 | 4531 | |||
| BC051954.1 | - | 3, 4 | 443 | 5714977 | ||
| BC060081.1 | - | 3, 4 | 443 | 5714977 | ||
| BC116751.1 | - | 4, 5 | 458 | 40126070 | ||
| BC116779.1 | - | 4, 5 | 458 | |||

Fam69b
Genomic Map