siRNA Details
| Gene Symbol: Ctsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019861.1 | NP_063914.1 | 4, 5 | 612 | ||
| GenBank mRNA | AF136280.1 | - | 4, 5 | 513 | ||
| AF197480.1 | - | 4, 5 | 557 | |||
| AJ131851.1 | - | 4, 5 | 489 | |||
| AK017474.1 | - | 4, 5 | 498 | |||
| AK075862.1 | - | 4, 5 | 505 | |||
| AK165416.1 | - | 3, 4 | 672 | |||
| BC004054.1 | - | 1, 2 | 83 | |||
| BC051131.1 | - | 3, 4 | 731 | |||
| BC058758.1 | - | 4, 5 | 503 | 6492834 | ||

Ctsf
Genomic Map