siRNA Details
| Gene Symbol: Zbtb20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285805.1 | NP_001272734.1 | 10 | 1710 | ||
| NM_019778.2 | NP_062752.2 | 8 | 1657 | |||
| NM_181058.1 | NP_851401.1 | 6 | 1435 | |||
| GenBank mRNA | AB221632.1 | - | 2 | 1122 | ||
| AF185576.1 | - | 6 | 1480 | |||
| AF194030.1 | - | 4 | 1272 | |||
| AK005028.2 | - | 6 | 1436 | |||
| AK034574.1 | - | 1 | 309 | |||
| AY028963.1 | - | 3 | 1239 | |||
| BC031114.1 | - | 7 | 1543 | 4971586 | ||
| BC056446.1 | - | 10 | 1682 | 5703319 | ||

Zbtb20
Genomic Map