siRNA Details
| Gene Symbol: Tex21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159532.1 | NP_001153004.1 | 4 | 721 | ||
| NM_019784.5 | NP_062758.3 | 4 | 721 | |||
| GenBank mRNA | AB000682.1 | - | 4 | 718 | ||
| AK016434.1 | - | 4 | 753 | |||
| AK016448.1 | - | 4 | 711 | |||
| AK016471.1 | - | 4 | 692 | |||
| BC100432.1 | - | 4 | 721 | 30917866 | ||

Tex21
Genomic Map