siRNA Details
| Gene Symbol: Bloc1s6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019788.3 | NP_062762.1 | 5 | 1644 | ||
| GenBank mRNA | AK038381.1 | - | 5 | 1622 | ||
| AK077720.1 | - | 5 | 1626 | |||
| AK147116.1 | - | 5 | 1645 | |||
| AK206166.1 | - | 1 | 121 | |||

Bloc1s6
Genomic Map