siRNA Details
| Gene Symbol: Ube2g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019803.3 | NP_062777.2 | 6 | 1468 | ||
| GenBank mRNA | AK003387.1 | - | 1 | 305 | ||
| AK136365.1 | - | 6 | 1458 | |||
| AK146554.1 | - | 6 | 1473 | |||
| AK154464.1 | - | 6 | 1489 | |||
| BC010321.1 | - | 6 | 1468 | 2651046 | ||

Ube2g2
Genomic Map