siRNA Details
| Gene Symbol: Anapc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019805.4 | NP_062779.3 | 8, 9 | 1274 | ||
| GenBank mRNA | AF076607.2 | - | 8, 9 | 1238 | ||
| AK030775.1 | - | 8, 9 | 1239 | |||
| AK049968.1 | - | 8, 9 | 1213 | |||
| AK137286.1 | - | 8, 9 | 1238 | |||
| AK150764.1 | - | 8, 9 | 1238 | |||
| AK150891.1 | - | 8, 9 | 1238 | |||
| AK153114.1 | - | 8, 9 | 1238 | |||
| AK159523.1 | - | 8, 9 | 1206 | |||
| AK164202.1 | - | 8, 9 | 1243 | |||
| BC006635.1 | - | 8, 9 | 1227 | 3482261 | ||

Anapc7
Genomic Map