siRNA Details
| Gene Symbol: Nudt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291046.1 | NP_001277975.1 | 5 | 1531 | ||
| NM_019837.2 | NP_062811.1 | 5 | 1523 | |||
| GenBank mRNA | AF264064.1 | - | 5 | 1522 | ||
| AK003601.1 | - | 1 | 514 | |||
| AK079658.1 | - | 3 | 1019 | |||
| BC016534.1 | - | 6 | 1430 | 2615904 | ||
| BC046805.1 | - | 5 | 1523 | 6829170 | ||

Nudt3
Genomic Map