siRNA Details
| Gene Symbol: Ctsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019861.1 | NP_063914.1 | 5 | 645 | ||
| GenBank mRNA | AF136280.1 | - | 5 | 546 | ||
| AF197480.1 | - | 5 | 590 | |||
| AJ131851.1 | - | 5 | 522 | |||
| AK017474.1 | - | 5 | 531 | |||
| AK075862.1 | - | 5 | 538 | |||
| AK165416.1 | - | 4 | 705 | |||
| BC004054.1 | - | 2 | 116 | |||
| BC051131.1 | - | 4 | 764 | |||
| BC058758.1 | - | 5 | 536 | 6492834 | ||

Ctsf
Genomic Map