siRNA Details
Gene Symbol: Ube2d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019912.2 | NP_064296.1 | 2 | 508 | ||
GenBank mRNA | AK005355.1 | - | 3 | 460 | ||
AK136197.1 | - | 2 | 446 | |||
AK139623.1 | - | 2 | 474 | |||
AK146659.1 | - | 2 | 481 | |||
AK147568.1 | - | 1 | 6194 | |||
AK148089.1 | - | 2 | 330 | |||
AK160567.1 | - | 2 | 481 | |||
AK162093.1 | - | 2 | 313 | |||
AK172088.1 | - | 2 | 335 | |||
AK190235.1 | - | 2 | 105 | |||
AK196425.1 | - | 2 | 105 | |||
AK199554.1 | - | 2 | 105 | |||
AK204863.1 | - | 2 | 106 | |||
AK219213.1 | - | 2 | 105 | |||
BC003923.1 | - | 2 | 489 | 3485259 | ||
U62483.1 | - | 2 | 464 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001037292, U13176.
Panther Classification:
Gene Ontology Categories:
