siRNA Details
| Gene Symbol: Mllt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019914.4 | NP_064298.1 | 2 | 882 | ||
| GenBank mRNA | AB000733.1 | - | 2 | 456 | ||
| AK046005.1 | - | 2 | 387 | |||
| AK152708.1 | - | 2 | 882 | |||
| AK158495.1 | - | 2 | 508 | |||
| BC022963.1 | - | 2 | 323 | 5365628 | ||
| U95498.1 | - | 2 | 443 | |||

Mllt11
Genomic Map