siRNA Details
| Gene Symbol: Mgst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019946.4 | NP_064330.2 | 2, 3 | 222 | ||
| GenBank mRNA | AF159050.1 | - | 1, 2 | 121 | ||
| AK002800.1 | - | 2, 3 | 223 | |||
| AK005122.1 | - | 2, 3 | 223 | |||
| BC009155.1 | - | 1, 2 | 161 | 2655738 | ||

Mgst1
Genomic Map