siRNA Details
| Gene Symbol: Stat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019963.1 | NP_064347.1 | 3 | 330 | ||
| GenBank mRNA | AF088862.1 | - | 3 | 200 | ||
| AF206162.1 | - | 3 | 330 | |||
| AK149838.1 | - | 3 | 327 | |||
| AK149924.1 | - | 3 | 343 | |||
| AK150070.1 | - | 3 | 342 | |||

Stat2
Genomic Map