siRNA Details
| Gene Symbol: Stap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019992.3 | NP_064376.1 | 1 | 106 | ||
| GenBank mRNA | AB036058.1 | - | 1 | 96 | ||
| AK041474.1 | - | 1 | 107 | |||
| AK042475.1 | - | 1 | 153 | |||
| AK153661.1 | - | 1 | 185 | |||
| AK162655.1 | - | 1 | 188 | |||
| BC057898.1 | - | 1 | 160 | |||
| BC145550.1 | - | 1 | 122 | 9054114 | ||
| BC145828.1 | - | 1 | 121 | 40131131 | ||
| BC145830.1 | - | 1 | 122 | 40131133 | ||

Stap1
Genomic Map