siRNA Details
| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019995.3 | NP_064379.1 | 4 | 271 | ||
| GenBank mRNA | AF139179.1 | - | 4 | 275 | ||
| AK050619.1 | - | 5 | 298 | |||
| AK083379.1 | - | 5 | 298 | |||
| AK210270.1 | - | 1 | 58 | |||
| AK212629.1 | - | 3 | 128 | |||
| BC052702.1 | - | 4 | 263 | 6400922 | ||

Supt20
Genomic Map