siRNA Details
| Gene Symbol: Sult1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019878.4 | NP_063931.1 | 2 | 226 | ||
| GenBank mRNA | AF022894.1 | - | 2 | 245 | ||
| AK033498.1 | - | 2 | 229 | |||
| AK087595.1 | - | 2 | 262 | |||
| BC024361.1 | - | 1 | 185 | 4990345 | ||
| U92076.1 | - | 2 | 171 | |||

Sult1b1
Genomic Map