siRNA Details
| Gene Symbol: Mpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164733.1 | NP_001158205.1 | 5 | 572 | ||
| NM_001164734.1 | NP_001158206.1 | 5 | 582 | |||
| NM_019939.2 | NP_064323.2 | 5 | 582 | |||
| GenBank mRNA | AF161181.1 | - | 5 | 446 | ||
| AF199009.1 | - | 5 | 578 | |||
| AF199010.1 | - | 5 | 578 | |||
| AK144434.1 | - | 5 | 583 | |||
| AK164549.1 | - | 5 | 573 | |||
| BC042868.2 | - | 5 | 529 | |||
| BC046421.1 | - | 5 | 529 | |||
| BC069908.1 | - | 5 | 571 | |||
| BC106140.1 | - | 5 | 529 | |||
| BC138666.1 | - | 5 | 440 | 8861687 | ||
| BC145363.1 | - | 5 | 440 | 9053921 | ||
| BC145364.1 | - | 5 | 430 | 9053922 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001134982, XM_006236496, XM_008762897, XM_008762895, XM_008762898, and others.

Mpp6
Genomic Map